Learn More
Cayman Chemical Fosfomycin (calcium salt), Purity: ≥95%, CAS: 26016-98-8, 1 g
SDP
Catalog No. 502913725
Encompass_Preferred
A broad-spectrum antibiotic; active against Salmonella and P. aeruginosa at 1.56 and 6.25 µg/ml, respectively, as well as MSSA, cephalosporin- and penicillin-resistant S. pneumoniae, MRSA, and vancomycin-resistant Enterococcus
Catalog No. 50-291-3725 Supplier Cayman Chemical Supplier No. 355351 G
A broad-spectrum antibiotic; active against Salmonella and P. aeruginosa at 1.56 and 6.25 µg/ml, respectively, as well as MSSA, cephalosporin- and penicillin-resistant S. pneumoniae, MRSA, and vancomycin-resistant Enterococcus