Learn More
Cayman Chemical FosfomycIncalcIum salt 25g
SDP
Catalog No. 502910143
Encompass_Preferred
A broad-spectrum antibiotic; active against Salmonella and P. aeruginosa at 1.56 and 6.25 UG/ml, respectively, as well as MSSA, cephalosporin- and penicillin-resistant S. pneumoniae, MRSA, and vancomycin-resistant Enterococcus
Catalog No. 50-291-0143 Supplier Cayman Chemical Supplier No. 3553525 G
A broad-spectrum antibiotic; active against Salmonella and P. aeruginosa at 1.56 and 6.25 UG/ml, respectively, as well as MSSA, cephalosporin- and penicillin-resistant S. pneumoniae, MRSA, and vancomycin-resistant Enterococcus