Learn More
Apexbio Technology LLC FOSFRUCTOSE-TRISODIUM-200MG
SDP
Catalog No. 5000568394
Encompass_Preferred
Fosfructose trisodium (CAS No 38099-82-0) is a compound studied in the field of biomedical research primarily for its role in energy metabolism pathways Originating from biochemical applications it influences fructose metabolism by acting on glycolysis and gluconeogenesis pathways potentially modifying cellular energy balance In vitro studies have demonstrated activity in the nanomolar to low micromolar range highlighting its potency in modulating metabolic enzymes Its typical applications include investigations using cell models and animal studies to determine its effects on metabolic disorders and it is also utilized in drug screening processes for targeting metabolic pathways Experimental concentrations typically depend on the specific research design adapting to the precise requirements of each study to elucidate its effects comprehensively
Catalog No. 50-005-68394 Supplier Apexbio Technology LLC Supplier No. C7025200
Fosfructose trisodium (CAS No 38099-82-0) is a compound studied in the field of biomedical research primarily for its role in energy metabolism pathways Originating from biochemical applications it influences fructose metabolism by acting on glycolysis and gluconeogenesis pathways potentially modifying cellular energy balance In vitro studies have demonstrated activity in the nanomolar to low micromolar range highlighting its potency in modulating metabolic enzymes Its typical applications include investigations using cell models and animal studies to determine its effects on metabolic disorders and it is also utilized in drug screening processes for targeting metabolic pathways Experimental concentrations typically depend on the specific research design adapting to the precise requirements of each study to elucidate its effects comprehensively