Learn More
Medchemexpress LLC HY-109115 5mg Medchemexpress, Fosifloxuridine nafalbenamide CAS:1332837-31-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1091155MG
Encompass_Preferred
Medchemexpress, HY-109115 5mg Fosifloxuridine nafalbenamide CAS:1332837-31-6 Fosifloxuridine nafalbenamide (NUC-3373), a pyrimidine nucleotide analogue, is a Thymidylate synthase inhibitor. Fosifloxuridine nafalbenamide is a ProTide transformation of fluorodeoxyuridine monophosphate. Fosifloxuridine nafalbenamide has anticancer activity. Fosifloxuridine nafalbenamide has the potential to evoke a host immune response and enhance immunotherapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0764