Learn More
Fosinopril Impurity A, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| Flash Point | Not Applicable |
| Molecular Formula | C23H34NO5P |
| Linear Formula | C23H34NO5P |
| Quantity | 25 mg |
| Synonym | (2 S,4 S)-4-Cyclohexyl-1-[[(R)-hydroxy(4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylic acid |
| Formula Weight | 435.49 |
| Grade | BP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.