Learn More
Medchemexpress LLC Fosinopril sodium | 88889-14-9 | 99.2% | 1 ML
SDP
Catalog No. 5000406396
Encompass_Preferred
Fosinopril sodium is an ester prodrug of an angiotensin-converting enzyme (ACE) inhibitor. It is used for the treatment of hypertension and some types of chronic heart failure. This product is an analytical standard intended for research and analytical applications. It has a purity of 99.2%.
  • Ester prodrug of an angiotensin-converting enzyme (ACE) inhibitor
  • Treats hypertension
  • Treats some types of chronic heart failure
  • Intended for research and analytical applications
  • Purity of 99.2%
Catalog No. 50-004-06396 Supplier Medchemexpress LLC Supplier No. HYB038210MM/1ML
Fosinopril sodium is an ester prodrug of an angiotensin-converting enzyme (ACE) inhibitor. It is used for the treatment of hypertension and some types of chronic heart failure. This product is an analytical standard intended for research and analytical applications. It has a purity of 99.2%.
  • Ester prodrug of an angiotensin-converting enzyme (ACE) inhibitor
  • Treats hypertension
  • Treats some types of chronic heart failure
  • Intended for research and analytical applications
  • Purity of 99.2%