Learn More
Cayman Chemical Fosinopril (sodium salt), Purity: ≥98%, CAS: 88889-14-9, 100 mg
SDP
Catalog No. 502985770
Encompass_Preferred
A prodrug form of fosinoprilat; inhibits angiotensin I-induced pressor responses in normotensive rats, dogs, and monkeys at 15, 15, and 10 µmol/kg, respectively; reduces fractional shortening and decreases left ventricular size in a porcine model of congestive heart failure at 2.5 mg/kg
Catalog No. 50-298-5770 Supplier Cayman Chemical Supplier No. 21899100 MG
A prodrug form of fosinoprilat; inhibits angiotensin I-induced pressor responses in normotensive rats, dogs, and monkeys at 15, 15, and 10 µmol/kg, respectively; reduces fractional shortening and decreases left ventricular size in a porcine model of congestive heart failure at 2.5 mg/kg