Learn More
Cayman Chemical FosInoprIlsodIum salt 50mg
SDP
Catalog No. 502987098
Encompass_Preferred
A prodrug form of fosinoprilat; inhibits angiotensin I-induced pressor responses in normotensive rats, dogs, and monkeys at 15, 15, and 10 µmol/kg, respectively; reduces fractional shortening and decreases left ventricular size in a porcine model of congestive heart failure at 2.5 mg/kg
Catalog No. 50-298-7098 Supplier Cayman Chemical Supplier No. 2189950 MG
A prodrug form of fosinoprilat; inhibits angiotensin I-induced pressor responses in normotensive rats, dogs, and monkeys at 15, 15, and 10 µmol/kg, respectively; reduces fractional shortening and decreases left ventricular size in a porcine model of congestive heart failure at 2.5 mg/kg