Learn More
Aviva Systems Biology Corp FOSL1 Antibody - middle region (ARP31378_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31378_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.SL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FOSL1. Alternate names: FRA1, fra-1, FRAApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54201