Learn More
Medchemexpress LLC Fosmidomycin sodium salt | 66508-37-0 | 99.7% | 205.08 | 10 MG
SDP
Catalog No. 5000437893
Encompass_Preferred
Fosmidomycin sodium salt is a phosphonic acid antibiotic and an antimalarial agent. It is active against both Gram-negative and Gram-positive bacteria.
  • Phosphonic acid antibiotic
  • Antimalarial agent
  • Active against Gram-negative bacteria
  • Active against Gram-positive bacteria
Catalog No. 50-004-37893 Supplier Medchemexpress LLC Supplier No. HY11285310MG
Fosmidomycin sodium salt is a phosphonic acid antibiotic and an antimalarial agent. It is active against both Gram-negative and Gram-positive bacteria.
  • Phosphonic acid antibiotic
  • Antimalarial agent
  • Active against Gram-negative bacteria
  • Active against Gram-positive bacteria