Learn More
Medchemexpress LLC Fosmidomycin sodium salt | 66508-37-0 | 99.7% | 205.08 | 100 MG
SDP
Catalog No. 5000437863
Encompass_Preferred
Fosmidomycin sodium salt is a phosphonic acid antibiotic and an antimalarial agent. It is active against both Gram-negative and Gram-positive bacteria.
  • Active against Gram-negative and Gram-positive bacteria.
  • Demonstrated efficacy as an antimalarial agent in animal models and clinical trials.
  • Effectively eliminates parasites in treated animals.
  • Reduces parasitemias to less than 1% at specified dosages.
  • Cures mice of malaria under specific treatment conditions.
Catalog No. 50-004-37863 Supplier Medchemexpress LLC Supplier No. HY112853100MG
Fosmidomycin sodium salt is a phosphonic acid antibiotic and an antimalarial agent. It is active against both Gram-negative and Gram-positive bacteria.
  • Active against Gram-negative and Gram-positive bacteria.
  • Demonstrated efficacy as an antimalarial agent in animal models and clinical trials.
  • Effectively eliminates parasites in treated animals.
  • Reduces parasitemias to less than 1% at specified dosages.
  • Cures mice of malaria under specific treatment conditions.