Learn More
Medchemexpress LLC Fosmidomycin sodium salt | 66508-37-0 | 99.7% | 205.08 | 50 MG
Supplier: Medchemexpress LLC HY11285350MG
Fosmidomycin sodium salt is a phosphonic acid antibiotic and an antimalarial agent, active against both Gram-negative and Gram-positive bacteria. It has shown effectiveness in in vivo studies, with animals treated intraperitoneally or orally becoming apparently free of parasites. Mice treated for 8 days were totally cured.
- Phosphonic acid antibiotic.
- Antimalarial agent.
- Active against both Gram-negative and Gram-positive bacteria.
- In vivo studies show that animals treated intraperitoneally with dosages >10 mg/kg are apparently free of parasites.
- In vivo studies show that animals treated orally with 50 or 100 mg/kg are apparently free of parasites.
- Mice treated with 30 mg/kg over an 8-day period were totally cured.