Learn More
Cayman Chemical FosmIdomycInsodIum salt 10mg
SDP
Catalog No. 502968591
Encompass_Preferred
An antibiotic that inhibits DOXP reductoisomerase, a key enzyme in the non-mevalonate pathway of isoprenoid biosynthesis; against both Gram-negative and Gram-positive bacteria and the human malarial parasite P. falciparum (IC50 = 290-370 nM)
Catalog No. 50-296-8591 Supplier Cayman Chemical Supplier No. 1600010 MG
An antibiotic that inhibits DOXP reductoisomerase, a key enzyme in the non-mevalonate pathway of isoprenoid biosynthesis; against both Gram-negative and Gram-positive bacteria and the human malarial parasite P. falciparum (IC50 = 290-370 nM)