Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fosphenytoin disodium | 92134-98-0 | MFCD20527322 | >98.0% | 406.24 g/mol | C16H13N2Na2O6P | 10 MG
SDP

Catalog No. 5000297815
Encompass_Preferred

Fosphenytoin disodium is the water-soluble disodium salt prodrug of phenytoin supplied for research use only. It functions as an anticonvulsant by reducing repetitive neuronal firing through blockade of voltage-gated sodium channels and is used as a reference compound in pharmacology and analytical studies.

  • water-soluble prodrug of phenytoin suitable for biochemical and pharmacological assays.
  • high purity (greater than 98%) for reliable experimental results.
  • molecular formula C16H13N2Na2O6P and molecular weight approximately 406.24 g/mol.
  • provided as a small research pack for analytical standards and in vitro testing.
  • intended for research use only; not for human or veterinary use.

Catalog No. 50-002-97815 Supplier Medchemexpress LLC Supplier No. HYB1657A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fosphenytoin disodium is the water-soluble disodium salt prodrug of phenytoin supplied for research use only. It functions as an anticonvulsant by reducing repetitive neuronal firing through blockade of voltage-gated sodium channels and is used as a reference compound in pharmacology and analytical studies.

  • water-soluble prodrug of phenytoin suitable for biochemical and pharmacological assays.
  • high purity (greater than 98%) for reliable experimental results.
  • molecular formula C16H13N2Na2O6P and molecular weight approximately 406.24 g/mol.
  • provided as a small research pack for analytical standards and in vitro testing.
  • intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.