Learn More
Cayman Chemical Fosphenytoin (sodium salt), Purity: ≥98%, CAS: 92134-98-0, 100 mg
SDP
Catalog No. 502995013
Encompass_Preferred
A prodrug of phenytoin; induces less tissue damage compared with two other phenytoin prodrugs in rats following s.c. and i.m. administration; has anticonvulsant activity in a mouse model of maximal electroshock seizures similar to that of phenytoin (ED50s = 16 and 8 mg/kg, respectively); has antiarrhythmic activity in isolated guinea pig atria and in anesthetized dogs
Catalog No. 50-299-5013 Supplier Cayman Chemical Supplier No. 23889100 MG
A prodrug of phenytoin; induces less tissue damage compared with two other phenytoin prodrugs in rats following s.c. and i.m. administration; has anticonvulsant activity in a mouse model of maximal electroshock seizures similar to that of phenytoin (ED50s = 16 and 8 mg/kg, respectively); has antiarrhythmic activity in isolated guinea pig atria and in anesthetized dogs