Learn More
Cayman Chemical FosphenytoInsodIum salt 500mg
SDP
Catalog No. 502994499
Encompass_Preferred
A prodrug of phenytoin; induces less tissue damage compared with two other phenytoin prodrugs in rats following s.c. and i.m. administration; has anticonvulsant activity in a mouse model of maximal electroshock seizures similar to that of phenytoin (ED50s = 16 and 8 mg/kg, respectively); has antiarrhythmic activity in isolated guinea pig atria and in anesthetized dogs
Catalog No. 50-299-4499 Supplier Cayman Chemical Supplier No. 23889500 MG
A prodrug of phenytoin; induces less tissue damage compared with two other phenytoin prodrugs in rats following s.c. and i.m. administration; has anticonvulsant activity in a mouse model of maximal electroshock seizures similar to that of phenytoin (ED50s = 16 and 8 mg/kg, respectively); has antiarrhythmic activity in isolated guinea pig atria and in anesthetized dogs