Learn More
Medchemexpress LLC HY-16779B 5mg Medchemexpress, Fosravuconazole (L-lysine ethanolate) CAS:914361-45-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16779B5MG
Encompass_Preferred
Medchemexpress, HY-16779B 5mg Fosravuconazole (L-lysine ethanolate) CAS:914361-45-8 Fosravuconazole L-lysine ethanolate (BMS-379224 L-lysine ethanolate), a prodrug of Ravuconazole, is an orally active broad spectrum antifungal agent. Fosravuconazole L-lysine ethanolate can be used for candidiasis, onychomycosis and parasitemia research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1911