Learn More
Medchemexpress LLC Fosravuconazole (L-lysine ethanolate) | 914361-45-8 | 99.9% | 739.73 | 25 MG
SDP
Catalog No. 5000391515
Encompass_Preferred
Fosravuconazole L-lysine ethanolate, a proagent of Ravuconazole, is an orally active broad spectrum antifungal agent. It can be used for candidiasis, onychomycosis, and parasitemia research.
  • Proagent of Ravuconazole
  • Orally active broad spectrum antifungal agent
  • Used for candidiasis research
  • Used for onychomycosis research
  • Used for parasitemia research
  • Potent in vitro antifungal activity against a wide range of fungal species, including Candida, Aspergillus, and Trichophyton species
  • Suppresses parasitemia and prevents death in mice infected with the T. cruzi Y strain (In Vivo data)
Catalog No. 50-003-91515 Supplier Medchemexpress LLC Supplier No. 25MGHY16779B
Fosravuconazole L-lysine ethanolate, a proagent of Ravuconazole, is an orally active broad spectrum antifungal agent. It can be used for candidiasis, onychomycosis, and parasitemia research.
  • Proagent of Ravuconazole
  • Orally active broad spectrum antifungal agent
  • Used for candidiasis research
  • Used for onychomycosis research
  • Used for parasitemia research
  • Potent in vitro antifungal activity against a wide range of fungal species, including Candida, Aspergillus, and Trichophyton species
  • Suppresses parasitemia and prevents death in mice infected with the T. cruzi Y strain (In Vivo data)