Learn More
Cayman Chemical FosravuconazoleLlysIn eth 10mg
SDP
Catalog No. 502944431
Encompass_Preferred
A prodrug form of ravuconazole; increases survival in a mouse model of systemic C. albicans infection
Catalog No. 50-294-4431 Supplier Cayman Chemical Supplier No. 4213310 MG
A prodrug form of ravuconazole; increases survival in a mouse model of systemic C. albicans infection