Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foss™ Soxtec™ 8000 Extraction Systems for Extractable Matter

Catalog No. TCS8000800G
Change view
Click to view available options
6 position, 1 basic unit, 1 control unit, 1 set of accessories
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
TCS8000800G 6 position, 1 basic unit, 1 control unit, 1 set of accessories
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TCS8000800G Supplier Foss™ Supplier No. S8000800G

missing translation for 'validMainframeMsg'

Includes

Accessories Include: 3 sets glass extraction cups 6/set, 1 set condenser seals, 2 thimble stands, 2 set thimble adapters 6/set, docking tool for thimbles, set of cellulose thimbles 33mm double thickness 25/set, thimble support, cup holder, cup stand, cup tool,cotton, solvent addition tube, solvent recovery flask, owners guide, application note, user manual, quick guide, spare parts manual

For fast and safe extraction of soluble matter from wide range of matrices

  • Fully automatic for unattended operation
  • Consists of basic unit and control unit, glass cups included
  • 6 hot plate positions, expandable up to 12
  • Sample size: 0.5 to 15g, range is application dependent
  • 100 to 120VAC or 200 to 240VAC, depending on version, 50/60 Hz
  • Power: 1500w. (120VAC version), 1800w. (230VAC version)
  • D x W x H: 350 x 640 x 630mm (extraction unit)

  • Closed solvent addition limits operator's exposure to solvent
  • Extraction cups use 70-90mL solvent, 80% of which is recovered
  • No touching or mixing of samples is ensured with the batch handling tools
  • Cooling water control reduces water costs
  • Flexible operation
  • Safe and consistent solvent dosage
  • Consistent solvent volume

Certifications

CE marked

Specifications

Includes 6 Position, One Basic Unit, One Control Unit, One set of Accessories; (Accessories Include: 3 sets glass extraction cups 6/set, 1 set condenser seals, 2 thimble stands, 2 set thimble adapters 6/set, docking tool for thimbles, set of cellulose thimbles 33mm double thickness 25/set, thimble support, cup holder, cup stand, cup tool,cotton, solvent addition tube, solvent recovery flask, owners guide, application note, user manual, quick guide, spare parts manual)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.