Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Fossils: How Do They Form?
SCI_ED

Catalog No. S02017 Shop All Aldon Products
Change view
Click to view available options
Grade:
K to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02017 K to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING CHOKING HAZARD Small parts. Not for children under 3 yrs.
Catalog No. S02017 Supplier Innovating Science™ Supplier No. IS1009
Add to Cart
Add to Cart
WARNING CHOKING HAZARD Small parts. Not for children under 3 yrs.

Teach students about the importance of fossils and how they form with a fun hands-on activity that simulates the formation of cast-and-mold fossils.

Teach students about the importance of fossils and how they form with a fun hands-on activity that simulates the formation of cast-and-mold fossils. Contains enough materials for 15 groups of students, Teacher's Manual, and Student Study Guide copymaster.

Specifications

Grade K to 5
Format Class Set
Product Type Fossil Kit
Languages English
Subject Geology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.