Learn More
Medchemexpress LLC HY-13038A 10mg , Fostamatinib CAS:901119-35-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13038A10MG
Encompass_Preferred
HY-13038A 10mg Fostamatinib CAS: 901119-35-5 Fostamatinib (R788) is the oral prodrug of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9338