Learn More
Selleck Chemical LLC Fostamatinib (R788) disodium S2206-250mg
SDP
Catalog No. 503112683
Encompass_Preferred
Fostamatinib disodium (R788 Tamatinib Fosdium) a prodrug of the active metabolite R406 is a Syk inhibitor with IC50 of 41 nM in a cell-free assay strongly inhibits Syk but not Lyn 5-fold less potent to Flt3 Phase 3
Catalog No. 50-311-2683 Supplier Selleck Chemical LLC Supplier No. S2206250MG
Fostamatinib disodium (R788 Tamatinib Fosdium) a prodrug of the active metabolite R406 is a Syk inhibitor with IC50 of 41 nM in a cell-free assay strongly inhibits Syk but not Lyn 5-fold less potent to Flt3 Phase 3