Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fostamatinib | 901119-35-5 | 99.7% | 25 MG
SDP

Catalog No. 5000341437
Encompass_Preferred

Fostamatinib is an oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor. It also inhibits Lyn and Lck, playing a role in various cellular pathways. This product is a solid form of Fostamatinib with high purity, suitable for laboratory research.

  • Oral proagent of active compound R406
  • Competitive Syk/FLT3 inhibitor
  • Also inhibits Lyn and Lck
  • High purity of 99.7%
  • Recommended storage for powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in-solvent at -80°C for 1 year or -20°C for 6 months
  • Shipped at room temperature for less than 2 weeks

Catalog No. 50-003-41437 Supplier Medchemexpress LLC Supplier No. 25MGHY13038A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fostamatinib is an oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor. It also inhibits Lyn and Lck, playing a role in various cellular pathways. This product is a solid form of Fostamatinib with high purity, suitable for laboratory research.

  • Oral proagent of active compound R406
  • Competitive Syk/FLT3 inhibitor
  • Also inhibits Lyn and Lck
  • High purity of 99.7%
  • Recommended storage for powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in-solvent at -80°C for 1 year or -20°C for 6 months
  • Shipped at room temperature for less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.