Learn More
Medchemexpress LLC Fostamatinib | 901119-35-5 | >99.7% | 50 MG
SDP
Catalog No. 5000340448
Encompass_Preferred
Fostamatinib (R788) is an orally available prodrug that acts as a potent and competitive Syk/FLT3 inhibitor. It also inhibits Lyn and Lck, making it a valuable tool for various research applications and substance manufacture. This compound is provided with high purity and comprehensive technical documentation.
  • Oral proagent of the active compound R406
  • Potent and competitive Syk/FLT3 inhibitor
  • Also inhibits Lyn and Lck
  • Purity greater than 99.7%
  • Provided as a solid appearance
  • Technical documentation includes data sheet, COA, SDS, and handling instructions
  • Suitable for use as a laboratory chemical
Catalog No. 50-003-40448 Supplier Medchemexpress LLC Supplier No. HY13038A50MG
Fostamatinib (R788) is an orally available prodrug that acts as a potent and competitive Syk/FLT3 inhibitor. It also inhibits Lyn and Lck, making it a valuable tool for various research applications and substance manufacture. This compound is provided with high purity and comprehensive technical documentation.
  • Oral proagent of the active compound R406
  • Potent and competitive Syk/FLT3 inhibitor
  • Also inhibits Lyn and Lck
  • Purity greater than 99.7%
  • Provided as a solid appearance
  • Technical documentation includes data sheet, COA, SDS, and handling instructions
  • Suitable for use as a laboratory chemical