Learn More
Medchemexpress LLC Fostamatinib Disodium | 1025687-58-4 | 98.3% | 200 MG
SDP
Catalog No. 5000338403
Encompass_Preferred
Fostamatinib Disodium is an oral prodrug of the active compound R406. It acts as an orally available and competitive Syk/FLT3 inhibitor, also demonstrating inhibitory activity against Lyn and Lck. This compound is intended for research purposes only.
  • Oral prodrug of R406
  • Orally available
  • Competitive Syk/FLT3 inhibitor (Ki of 30 nM, IC50 of 41 nM)
  • Inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM)
  • Intended for research use only
Catalog No. 50-003-38403 Supplier Medchemexpress LLC Supplier No. HY13038200MG
Fostamatinib Disodium is an oral prodrug of the active compound R406. It acts as an orally available and competitive Syk/FLT3 inhibitor, also demonstrating inhibitory activity against Lyn and Lck. This compound is intended for research purposes only.
  • Oral prodrug of R406
  • Orally available
  • Competitive Syk/FLT3 inhibitor (Ki of 30 nM, IC50 of 41 nM)
  • Inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM)
  • Intended for research use only