Learn More
Medchemexpress LLC Fostamatinib Disodium | 1025687-58-4 | 98.3% | 25 MG
SDP
Catalog No. 5000342297
Encompass_Preferred
Fostamatinib Disodium is the oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).
  • Oral proagent of active compound R406
  • Orally available and competitive Syk/FLT3 inhibitor
  • Inhibits Lyn and Lck
Catalog No. 50-003-42297 Supplier Medchemexpress LLC Supplier No. 25MGHY13038
Fostamatinib Disodium is the oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).
  • Oral proagent of active compound R406
  • Orally available and competitive Syk/FLT3 inhibitor
  • Inhibits Lyn and Lck