Learn More
TARGETMOL CHEMICALS INC FOSTAMATINIB DISODIUM 10MG
SDP
Catalog No. 502577082
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fostamatinib Disodium (R788 Disodium) is an orally available Syk kinase inhibitor with potential anti-inflammatory and immunomodulating activities. purity: 98%
Catalog No. 50-257-7082 Supplier TARGETMOL CHEMICALS INC Supplier No. T260510MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fostamatinib Disodium (R788 Disodium) is an orally available Syk kinase inhibitor with potential anti-inflammatory and immunomodulating activities. purity: 98%