Learn More
Medchemexpress LLC HY-13038 5mg Medchemexpress, Fostamatinib Disodium CAS:1025687-58-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130385MG
Encompass_Preferred
Medchemexpress, HY-13038 5mg Fostamatinib Disodium CAS:1025687-58-4 Fostamatinib Disodium (R788 Disodium), a prodrug of the active metabolite R406, is a potent Syk inhibitor with IC50 of 41 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8953
 Restricted Item