Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fostamatinib (disodium hexahydrate) | 914295-16-2 | 99.5% | 100 MG
SDP

Catalog No. 5000338402
Encompass_Preferred

Fostamatinib (disodium hexahydrate) is a chemical compound commonly used as a laboratory chemical and for the manufacture of substances. It appears as a white to gray solid with a high purity of 99.5%. It is recommended for sealed storage away from moisture at 4°C, or at -80°C for 6 months (-20°C for 1 month) when in solvent.

  • Used in laboratory research
  • Suitable for substance manufacturing
  • High purity (99.5%)
  • Recommended sealed storage at 4°C, away from moisture
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks
  • White to gray solid appearance

Catalog No. 50-003-38402 Supplier Medchemexpress LLC Supplier No. HY13038B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fostamatinib (disodium hexahydrate) is a chemical compound commonly used as a laboratory chemical and for the manufacture of substances. It appears as a white to gray solid with a high purity of 99.5%. It is recommended for sealed storage away from moisture at 4°C, or at -80°C for 6 months (-20°C for 1 month) when in solvent.

  • Used in laboratory research
  • Suitable for substance manufacturing
  • High purity (99.5%)
  • Recommended sealed storage at 4°C, away from moisture
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks
  • White to gray solid appearance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.