Learn More
Medchemexpress LLC Fostamatinib (disodium hexahydrate) | 914295-16-2 | 99.5% | 25 MG
SDP
Catalog No. 5000341699
Encompass_Preferred
Fostamatinib (disodium hexahydrate) is a laboratory chemical provided as a solid with a purity of 99.5%. It requires sealed storage at 4°C away from moisture. When in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month.
  • Stable under recommended storage conditions
  • Requires sealed storage to protect from moisture
  • Recommended storage at 4°C
  • For solutions, store at -80°C (up to 6 months) or -20°C (up to 1 month)
Catalog No. 50-003-41699 Supplier Medchemexpress LLC Supplier No. 25MGHY13038B
Fostamatinib (disodium hexahydrate) is a laboratory chemical provided as a solid with a purity of 99.5%. It requires sealed storage at 4°C away from moisture. When in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month.
  • Stable under recommended storage conditions
  • Requires sealed storage to protect from moisture
  • Recommended storage at 4°C
  • For solutions, store at -80°C (up to 6 months) or -20°C (up to 1 month)