Learn More
Medchemexpress LLC Fostamatinib disodium hexahydrate | 914295-16-2 | 99.5% | 50 MG
SDP
Catalog No. 5000338326
Encompass_Preferred
Fostamatinib disodium hexahydrate is an oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor, also inhibiting Lyn and Lck. This product is for research use only.
  • Oral proagent of R406
  • Orally available
  • Competitive Syk/FLT3 inhibitor
  • Inhibits Lyn and Lck
  • For research use only
Catalog No. 50-003-38326 Supplier Medchemexpress LLC Supplier No. HY1303850MG
Fostamatinib disodium hexahydrate is an oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor, also inhibiting Lyn and Lck. This product is for research use only.
  • Oral proagent of R406
  • Orally available
  • Competitive Syk/FLT3 inhibitor
  • Inhibits Lyn and Lck
  • For research use only