Learn More
Medchemexpress LLC FOSTAMATINIB (R788) 5MG
SDP
Catalog No. NC1848513
Encompass_Preferred
Fostamatinib (R788), a prodrug of the active metabolite R406, is a potent Syk inhibitor with IC50 of 41 nM 99.20% CAS#901119-35-5 C₂₃H₂₆FN₆O₉P
Catalog No. NC1848513 Supplier Medchemexpress LLC Supplier No. HY13038A5MG
Fostamatinib (R788), a prodrug of the active metabolite R406, is a potent Syk inhibitor with IC50 of 41 nM 99.20% CAS#901119-35-5 C₂₃H₂₆FN₆O₉P