Learn More
Medchemexpress LLC HY-15440A 5mg Medchemexpress, Fostemsavir CAS:864953-29-7 Purity:>98%
SDP
Catalog No. 501932737
Encompass_Preferred
Medchemexpress, HY-15440A 5mg Fostemsavir CAS:864953-29-7 Fostemsavir (BMS-663068) is the phosphonooxymethyl prodrug of BMS-626529. Fostemsavir (BMS-663068) is a novel attachment inhibitor that targets HIV-1 gp120 and prevents its binding to CD4 + T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2737 Supplier Medchemexpress LLC Supplier No. HY15440A5MG
Medchemexpress, HY-15440A 5mg Fostemsavir CAS:864953-29-7 Fostemsavir (BMS-663068) is the phosphonooxymethyl prodrug of BMS-626529. Fostemsavir (BMS-663068) is a novel attachment inhibitor that targets HIV-1 gp120 and prevents its binding to CD4 + T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.