Learn More
Medchemexpress LLC Fostemsavir Tris | 864953-39-9 | C29H37N8O11P | 10 MM/ML
SDP
Catalog No. 5000376645
Encompass_Preferred
Fostemsavir Tris (BMS-663068 (Tris)) is a phosphonooxymethyl proagent of BMS-626529. This novel attachment inhibitor targets HIV-1 gp120, preventing its binding to CD4+ T cells. It demonstrates significant antiviral activity in subjects with susceptible viruses, making it a valuable compound for research applications.
  • Targets HIV-1 gp120
  • Prevents binding to CD4+ T cells
  • Exhibits good antiviral activity
  • Intended for research use
  • Available as a solution in DMSO
Catalog No. 50-003-76645 Supplier Medchemexpress LLC Supplier No. HY15440B10MM/1ML
Fostemsavir Tris (BMS-663068 (Tris)) is a phosphonooxymethyl proagent of BMS-626529. This novel attachment inhibitor targets HIV-1 gp120, preventing its binding to CD4+ T cells. It demonstrates significant antiviral activity in subjects with susceptible viruses, making it a valuable compound for research applications.
  • Targets HIV-1 gp120
  • Prevents binding to CD4+ T cells
  • Exhibits good antiviral activity
  • Intended for research use
  • Available as a solution in DMSO