Learn More
MSC Fostoria Electric Infrared Heater, 4.8 kW, 240 V, Mul-T-Mount, Stainless Steel
Catalog No. 5000507081
Encompass
This electric infrared heater is designed for indoor and outdoor commercial/industrial applications. With a durable stainless steel housing and gold anodized aluminum reflectors, this 4.8 kW, 240 V heater provides efficient, instantaneous heat for comfort heating, spot heating, and snow/ice control.
  • Durable stainless steel housing.
  • Gold anodized aluminum reflectors for superior reflectivity.
  • High moisture and thermal shock resistant elements; 96% radiant efficiency.
  • Instantaneous heat-up and cool-down.
  • High watt densities for effective heating.
  • Suitable for indoor/outdoor commercial/industrial use.
  • Built-in surface mounting bracket with 45° horizontal tilt.
  • Operates on single or three-phase wiring.
  • Provides high-intensity radiant infrared heating.
  • Supports staging and zone control.
Catalog No. 50-005-07081 Supplier MSC Supplier No. 76933423
This electric infrared heater is designed for indoor and outdoor commercial/industrial applications. With a durable stainless steel housing and gold anodized aluminum reflectors, this 4.8 kW, 240 V heater provides efficient, instantaneous heat for comfort heating, spot heating, and snow/ice control.
  • Durable stainless steel housing.
  • Gold anodized aluminum reflectors for superior reflectivity.
  • High moisture and thermal shock resistant elements; 96% radiant efficiency.
  • Instantaneous heat-up and cool-down.
  • High watt densities for effective heating.
  • Suitable for indoor/outdoor commercial/industrial use.
  • Built-in surface mounting bracket with 45° horizontal tilt.
  • Operates on single or three-phase wiring.
  • Provides high-intensity radiant infrared heating.
  • Supports staging and zone control.