Learn More
MSC Fostoria Infrared Quartz Electric Heater, 4800W, 277V, 1 ea
Catalog No. 5000506958
Encompass
The Fostoria Mul-T-Mount THSS Series Infrared Quartz Electric Heater is a 3-lamp unit designed for commercial and industrial heating applications. Operating at 4800W and 277V, it is suitable for both indoor and unprotected outdoor environments, including recessed installations. This robust heater utilizes Fostoria quartz lamp infrared elements to provide efficient spot or total building heating.
  • Three-lamp configuration with stainless steel housing
  • Equipped with 6 high-temperature silicone lead wires for single or three-phase service
  • Features a 'T' tab wire channel for simplified installation
  • Non-corrosive housing and hardware ensures durability in harsh conditions
  • Utilizes clear 12 mm diameter quartz lamps
  • Provides bright light, generating 8 lumens per watt
Catalog No. 50-005-06958 Supplier MSC Supplier No. 76933431
The Fostoria Mul-T-Mount THSS Series Infrared Quartz Electric Heater is a 3-lamp unit designed for commercial and industrial heating applications. Operating at 4800W and 277V, it is suitable for both indoor and unprotected outdoor environments, including recessed installations. This robust heater utilizes Fostoria quartz lamp infrared elements to provide efficient spot or total building heating.
  • Three-lamp configuration with stainless steel housing
  • Equipped with 6 high-temperature silicone lead wires for single or three-phase service
  • Features a 'T' tab wire channel for simplified installation
  • Non-corrosive housing and hardware ensures durability in harsh conditions
  • Utilizes clear 12 mm diameter quartz lamps
  • Provides bright light, generating 8 lumens per watt