Learn More
Cayman Chemical FostrIecInsodIum salt 100ug
SDP
Catalog No. 502970094
Encompass_Preferred
An inhibitor of PP2A and PP4 (IC50s = 3.2 and 3 nM, respectively); less effectively inhibits topoisomerase II and PP1 (IC50s = 40 and 131 μM, respectively) and does not inhibit PP2B
Catalog No. 50-297-0094 Supplier Cayman Chemical Supplier No. 14749100 UG
An inhibitor of PP2A and PP4 (IC50s = 3.2 and 3 nM, respectively); less effectively inhibits topoisomerase II and PP1 (IC50s = 40 and 131 μM, respectively) and does not inhibit PP2B