Learn More
Electron Microscopy Sciences Fouchets Reagent 250 ML
SDP
Catalog No. 5031863
Encompass_Preferred
Used in Hall's Method for bilirubin. References: Hall, M.J., Amer. J. Clin., 34:313 –316, 1960 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, N.Y., p. 174, c. 1968
Catalog No. 50-318-63 Supplier Electron Microscopy Sciences Supplier No. 2661101
Used in Hall's Method for bilirubin. References: Hall, M.J., Amer. J. Clin., 34:313 –316, 1960 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, N.Y., p. 174, c. 1968