Learn More
Chemglass Life Sciences 10mL Single Neck Round Bottom Flask (RBF) , 14/10 Threaded Outer Joint
SDP
Catalog No. MW5802
Encompass_Preferred
10mL Single Neck RBF, 14/10 Threaded Outer Joint. Micro round bottom flask with a 14/10 threaded outer joint. Supplied complete with an open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).
Catalog No. MW5802 Supplier Chemglass Life Sciences Supplier No. MW5802
10mL Single Neck RBF, 14/10 Threaded Outer Joint. Micro round bottom flask with a 14/10 threaded outer joint. Supplied complete with an open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).