Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio Foundation™ Fetal Bovine Serum
SDP

Catalog No. 11852141 Shop All Gemini Bio Products
Change view
Click to view available options
Grade:
Economy
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
11-852-141 Economy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-852-141 Supplier GeminiBio Supplier No. 900108500ML
Get 1 Extra Serum When You Buy 5
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cost-effective serum that delivers excellent cell growth at an economical price.

  • Cost-effective FBS for excellent cell growth at an economical price point
  • Derived from USDA-approved collection facilities
  • Same filtration process and final quality-control testing as GeminiBio premium grades; allows slightly higher hemoglobin and endotoxin levels
  • Triple 0.1 μm sterile-filtered for exceptional purity
  • Mycoplasma-tested and virus-screened
  • Up to 5-year shelf life from date of manufacture
  • Ships on dry ice; store frozen at -20°C to -10°C
  • Ideal for general cell culture, stem cell research, immunology, neuroscience, cancer research, and biotechnology
  • Manufactured under an ISO 13485:2016 certified quality system
  • Complimentary heat inactivation upon request

Specifications

Additional Assays Biochemical profile, Electrophoretic profile
Certifications/Compliance ISO 13485:2016 certified quality system
Content And Storage 500 mL bottle, store frozen at -20°C to -10°C
Species Bovine
Age Fetal
Country of Origin Countries meeting USDA importation requirements
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Fetal bovine blood
Grade Economy
Processing Triple 0.1 μm sterile-filtered
Tested For Mycoplasma, Sterility (bacteria and fungi), Adventitious viruses
Formulation Liquid, frozen
For Use With (Application) Cell Culture
Heat Inactivated No
Lyophilized No
Product Line Foundation™
Mycoplasma Detected No
Shipping Condition Dry Ice
Regulatory Status RUO
Gamma-irradiated No
Show More Show Less

For cell culture, further manufacturing or research use only. Not for direct therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.