Learn More
Grainger Kasco Decorative Fountain Pump, 1 hp, 120 V, Stainless Steel
Catalog No. 502789453
Encompass_Preferred
The 4400JF fountain is a 1 hp decorative floating fountain system designed for reliable, continuous-duty aeration and aesthetic display in ponds and other water features. This complete package is engineered for ease of use in shallow water environments, featuring a surface-intake design that enables operation at depths as low as 19 in. The unit is constructed with durable, corrosion-resistant materials suitable for both freshwater and saltwater applications and is safety-tested to UL and CSA standards.
  • Includes 5 interchangeable nozzle spray patterns for versatile water displays
  • Features a 1 hp, 120V motor with thermal overload protection
  • Comes with a 150 ft. SJTOW power cord and a rain-proof control center with a 15 amp GFCI breaker and 24-hour timer
  • Equipped with two 50 ft. braided nylon mooring ropes for secure positioning
  • Utilizes a unitized carbon-ceramic mechanical seal and sacrificial
Catalog No. 50-278-9453 Supplier Grainger Supplier No. 16A124
The 4400JF fountain is a 1 hp decorative floating fountain system designed for reliable, continuous-duty aeration and aesthetic display in ponds and other water features. This complete package is engineered for ease of use in shallow water environments, featuring a surface-intake design that enables operation at depths as low as 19 in. The unit is constructed with durable, corrosion-resistant materials suitable for both freshwater and saltwater applications and is safety-tested to UL and CSA standards.
  • Includes 5 interchangeable nozzle spray patterns for versatile water displays
  • Features a 1 hp, 120V motor with thermal overload protection
  • Comes with a 150 ft. SJTOW power cord and a rain-proof control center with a 15 amp GFCI breaker and 24-hour timer
  • Equipped with two 50 ft. braided nylon mooring ropes for secure positioning
  • Utilizes a unitized carbon-ceramic mechanical seal and sacrificial