Learn More
World Precision Instrument Four-Channel Free Radical Analyzer
SDP
Catalog No. 502521045
Encompass
Real-time detection and measurement of a variety of redox-reactive species is fast and easy using the electrochemical (amperometric) detection principle employed in the  TBR4100. This optically isolated four-channel free radical analyzer has ultra low noise and independently operated channels.
To learn more about our warranty options, click here.
Catalog No. 50-252-1045 Supplier World Precision Instrument Supplier No. TBR4100
Add to Cart
Add to Cart
Request a Quote
Real-time detection and measurement of a variety of redox-reactive species is fast and easy using the electrochemical (amperometric) detection principle employed in the  TBR4100. This optically isolated four-channel free radical analyzer has ultra low noise and independently operated channels.
To learn more about our warranty options, click here.