Learn More
Electron Microscopy Sciences Four Pin Stub Holder
SDP
Catalog No. 5026070
Encompass
It accommodates four pin type, up to 12.5 ( 1/2") surface specimen stubs, with 1/8" (3.1 mm) diameter pin.
Catalog No. 50-260-70 Supplier Electron Microscopy Sciences Supplier No. 7594404
It accommodates four pin type, up to 12.5 ( 1/2") surface specimen stubs, with 1/8" (3.1 mm) diameter pin.