Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laxco Four Quadrant LED Ring Light for Stereo Microscopes
GREENER_CHOICESDP

Catalog No. 12563735 Shop All Laxco Inc Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
12-563-735 100/115/220/230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-563-735 Supplier Laxco Supplier No. AMPSRLED4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ring Light for Stereo Microscopes, 144 LED array, White Light, 3 interchangable settings with controller (full/half/quarter), No shadow on sample. Compatible with LAXCO LMS series stereo microscope.

  • 144 bright white LED bulbs
  • 100,000 hours of life
  • Intense and focused shadow-free illumination
  • White Light, 3 interchangeable settings with controller
  • Variable intensity control using controller
  • Inside diameter of ring light: 2-1/2 in. (64 mm)
  • Outside diameter of ring light: 4 in. (100 mm)
  • Outside diameter of ring light: 4 in. (100 mm)
  • Power input: 110-240 V, 50-60 HZ, auto switching

Specifications

Type Fiber Optic Ringlight
Autoclavable Not Autoclavable
For Use With (Equipment) Stereo Microscopes
Voltage 100/115/220/230 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.