Learn More
Prior Scientific Inc Four Slide Holder, Wellplate Format, Designed for Inverted Microscopes
Catalog No. NC2752647
Encompass_Preferred
Maximum accepted slide dimensions: 76.5mm x 27mm x 1.1mmMinimum accepted slide dimensions: 75.2mm x 25mm x 0.9mmMaterial: Black polycarbonate
Catalog No. NC2752647 Supplier Prior Scientific Inc Supplier No. W4416
Maximum accepted slide dimensions: 76.5mm x 27mm x 1.1mmMinimum accepted slide dimensions: 75.2mm x 25mm x 0.9mmMaterial: Black polycarbonate