Learn More
Diversified Spaces™ Four-Station Steel Workbench with 12 Vertical Locker Openings
GSA_VASCI_ED
Catalog No. S42636 Shop All Diversified Spaces Products
Change view
Click to view available options
Color:
Black
Includes:
Four vises
No vises
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
S42636 Black Four vises
S42635 Black No vises
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S42636 Supplier Diversified Spaces™ Supplier No. WB12BL4V
Add to Cart
Add to Cart
Request a Quote
Double-faced heavy gauged steel unit with one section construction and reinforced side panels available with or without vises in gray or black colors.
  • Base consists of double-faced 20 gauge steel unit, welded and riveted throughout
  • Unit provides 12 vertical locker openings of 53.34D x 30.48W x 38.10cm H (21 x 12 x 15 in.) dimension
  • 18 gauge steel locker doors with air vents, spring hinges, padlock hasp and knockout plug for cylinder lock
  • Steel lockers with gray baked-enamel finish
  • Maple top with 5.715 (2.25 in.) thickness
  • SEFA compliant and MAS Certified Green
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type 4-Station Steel Workbench with 12 Vertical Locker Openingss
Dimensions (D x W x H) 137.16 x 162.56 x 84.450 cm (54 x 64 x 33.25 in.)
Warranty Limited Lifetime
Includes Four vises
Color Black
Material Metal