Learn More
Diversified Spaces™ Four-Station Table
SCI_ED
Catalog No. S42473 Shop All Diversified Spaces Products
Change view
Click to view available options
Product Type:
4-Station Table with Maple Top
4-Station Table with Plastic Laminate Top
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S42473 4-Station Table with Plastic Laminate Top
S42471 4-Station Table with Maple Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S42473 Supplier Diversified Spaces™ Supplier No. WX4PM
Add to Cart
Add to Cart
Designed to provide structurally sound and completely functional work center
  • Constructed from solid maple and maple veneer
  • 2.063 x 11.43cm (0.8125 x 4.5 in.) solid maple aprons with 13-gauge steel corner braces
  • 5.715cm (2.25 in.) maple legs with adjustable floor glides and rubber leg boots
  • SEFA compliant and MAS Certified Green
  • Finished with durable chemical resistant non-emitting UV finish
Specifications
Product Type 4-Station Table with Plastic Laminate Top
Thickness (English) 1.25 in.
Warranty Limited Lifetime
Color Almond
Dimensions (D x W x H) 121.92 x 121.92 x 76.2 cm (48 x 48 x 30 in.)
Thickness (Metric) 31.75 mm
Material Maple