Learn More
Cole-Parmer Fowler 54-100-444-1 Stainless Steel Digital Caliper w/RS-232; 0 to 6" (0 to 150mm)
Catalog No. NC1680438 Shop All Cole-Parmer Products
Encompass_Preferred
IP67 moisture-resistant; Extra large screen, ditial output; measure inside, outside, depth, step; direct inch/metric conversion; thumb roller, stainless steel construction.
Catalog No. NC1680438 Supplier Cole-Parmer Supplier No. 9715200
Add to Cart
Add to Cart
IP67 moisture-resistant; Extra large screen, ditial output; measure inside, outside, depth, step; direct inch/metric conversion; thumb roller, stainless steel construction.