Learn More
MSC Fowler Adjustable Parallel Set, 6 pc, 3/8-2-1/4 in Range
Catalog No. 5000537068
Encompass
This adjustable parallel set includes six parallels designed for precise measurement and setup in various applications. Each parallel features two dovetailed pieces that slide and lock into position with set screws, allowing for easy adjustment. They can be quickly inserted into slots and expanded to size, with height size ranges from 3/8 in to 2-1/4 in. The slot size is determined by external measurement with a micrometer.
  • Set of 6 adjustable parallels
  • Each parallel has two dovetailed pieces that slide on each other and lock in position with set screws
  • Easily adjusted and set up to size with a micrometer
  • Can be quickly inserted in slots and expanded to size
  • Height size ranges from 3/8 in to 2-1/4 in
  • Slot size determined by measuring the parallels externally with a micrometer
Catalog No. 50-005-37068 Supplier MSC Supplier No. 98268527
This adjustable parallel set includes six parallels designed for precise measurement and setup in various applications. Each parallel features two dovetailed pieces that slide and lock into position with set screws, allowing for easy adjustment. They can be quickly inserted into slots and expanded to size, with height size ranges from 3/8 in to 2-1/4 in. The slot size is determined by external measurement with a micrometer.
  • Set of 6 adjustable parallels
  • Each parallel has two dovetailed pieces that slide on each other and lock in position with set screws
  • Easily adjusted and set up to size with a micrometer
  • Can be quickly inserted in slots and expanded to size
  • Height size ranges from 3/8 in to 2-1/4 in
  • Slot size determined by measuring the parallels externally with a micrometer